Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APPBP2 Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 124091AP150UL
Description
APPBP2 Polyclonal antibody specifically detects APPBP2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
APPBP2 | |
Polyclonal | |
Unconjugated | |
Q92624 | |
Rabbit | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
APP-BP2; APPBP2; HS.84084; KIAA0228; PAT1 | |
Antigen affinity chromatography | |
RUO | |
10513 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction