Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ApoCIII Polyclonal Antibody, Bethyl Laboratories
Goat Polyclonal Antibody
$296.84 - $462.53
Specifications
Antigen | ApoCIII |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-264-0707
|
Bethyl Laboratories, Inc
A81118A500UG |
500 μg |
Each for $296.84
|
|
|||||
50-264-0708
|
Bethyl Laboratories, Inc
A81118A1MG |
1 mg |
Each for $462.53
|
|
|||||
Description
ApoCIII Polyclonal antibody specifically detects ApoCIII in Human samples. It is validated for ELISA, Western BlotSpecifications
ApoCIII | |
ELISA, Western Blot | |
Unconjugated | |
Goat | |
Human | |
P02656 | |
345 | |
Primary | |
4° C | |
APOC3 |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
75mM sodium phosphate with 0.5mM EDTA, 75mM NaCl and 0.02% sodium azide; pH 7.2 | |
Apoc3; APOCIII; Apo-CIII; apoC-III; Apolipoprotein; Apolipoprotein C3; apolipoprotein C-3; apolipoprotein C-III; HALP2; MGC150353 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title