Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ApoCII Polyclonal Antibody, Biotin, Bethyl Laboratories
Goat Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A81114B
Description
ApoCII Polyclonal antibody specifically detects ApoCII in Human samples. It is validated for ELISA, Western BlotSpecifications
ApoCII | |
Polyclonal | |
Biotin | |
APOC2 | |
APC 2; APC2; APO C2; Apo CII; APOC 2; ApoC II; Apoc2; APOC2 protein; ApoCII; APO-CII; apoC-II; Apolipoprotein C II; apolipoprotein C2; Apolipoprotein CII; Apolipoprotein C-II; ApolipoproteinCII; MGC75082; ProapoC-II; Proapolipoprotein C-II; RGD1560725 | |
Antigen affinity chromatography | |
RUO | |
344 | |
4° C | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
75mM sodium phosphate with 0.5mM EDTA, 75mM NaCl and 0.02% sodium azide; pH 7.2 | |
P02655 | |
Goat | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction