Learn More
Fluidigm Corp Anti-Mouse Ly-6C (HK1.4)-150Nd 25 Tests
SDP
Supplier:  Fluidigm Corp 3150010C
Encompass_Preferred
Ly-6C is a 14–17 kDa GPI-linked cell surface glycoprotein that is widely used to identify functionally discrete murine-circulating monocyte populations.
Catalog No. 50-196-2708
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.