Learn More
Fluidigm Corp Anti-Mouse CD39 (24DMS1)-142Nd 25 Tests
SDP
Supplier:  Fluidigm Corp 3142005C
Encompass_Preferred
CD39 is expressed in B cells, subsets of dendritic cells, most monocytes, and CD4+CD25+ immunosuppressive regulatory T (Treg) cells.
Catalog No. 50-196-2501
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.