Learn More
Fluidigm Corp Anti-Human CD23 (EBVCS-5)-164Dy 100 Tests
SDP
Supplier:  Fluidigm Corp 3164018B
Encompass_Preferred
CD23, the low-affinity receptor for immunoglobulin E, is expressed on monocytes, mantle zone B cells, mature B cells, and follicular dendritic cells and at low levels on T, NK, and Langerhans cells and platelets.
Catalog No. 50-196-3128
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.