Learn More
Fluidigm Corp Anti-Cross GFAP (GA5)-143Nd 100 Tests
Supplier: Fluidigm Corp 3143022B
Glial fibrillary acidic protein (GFAP) is a 49 kDa cytoskeletal type III intermediate filament and maintains cell stability, shape, motility and proliferation.