Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Serum, Porcine, Polyclonal, MP Biomedicals
Supplier: MP Biomedicals, Inc 08650351
Description
- Primary Antibodies
- Application: Immunoassays
- Host: Rabbit
- Polyclonal
- Antigen: Anti-Porcine Serum
- Host: Rabbit
- Form: Lyophilized
Specifications
Polyclonal | |
Unconjugated | |
2 mL | |
Porcine |
N/A | |
Rabbit | |
Primary |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction