Learn More
PBL Assay Science Anti-Mouse Interferon Beta (IFN-Beta) Rabbit Polyclonal Antibody (PAb) [2e4 (20,000) units/vial]
Supplier:  PBL Assay Science 324001
Encompass_Preferred
Rabbit polyclonal antibody Anti-Mouse Interferon Beta is manufactured by PBL Assay Science (Catalog #32400-1) for research use only (RUO) and packaged in 2e4 (20,000) units/vial. This PAb binds to and neutralizes mouse IFN beta. It is suitable for use in neutralization, immunohistochemistry, immunoprecipitation, ELISA, and Western Blot applications.
Catalog No. 50-153-8048
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.