Learn More
STEMCELL Technologies Anti-Mouse F4/80 Antibody, Clone BM8, FITC, 50 µg
SDP
Supplier:  STEMCELL Technologies 60027FI.1
Encompass_Preferred
Rat monoclonal IgG2a antibody against human, mouse F4/80, FITC-conjugated
Catalog No. 50-197-5135
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.