Learn More
Fluidigm Corp ANTI-MOUSE CD3E -152SM
SDP
Supplier:  Fluidigm Corp 3152004B
Encompass_Preferred
Mouse CD3ε is a 20 kDa subunit of the TCR complex. There are three CD3 subunits, epsilon, gamma, and delta, and they are all required for proper assembly, trafficking, and surface expression of the TCR complex. CD3 is expressed at different levels on thymocytes during T cell differentiation and by all mature T cells. Binding of 145-2C11 to TCR initiates the intracellular biochemical pathway resulting in cellular activation, proliferation, and apoptosis, depending on specific conditions utilized. 145-2C11 is commonly used as a phenotypic marker for mouse T cells.
Catalog No. 50-466-866
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.