Learn More
Fluidigm Corp ANTI-MOUSE CD127/IL7RA-175LU
SDP
Supplier:  Fluidigm Corp 3175006B
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CD127, also known as the IL-7 receptor α chain or IL-7α, is a 60–90 kDa type I transmembrane glycoprotein. It is a heterodimer consisting of IL-7Rα (CD127) and a common-γ chain receptor (CD132), which is shared with various cytokines including IL-2,-4,-9,-13,-15 and -21. CD127 is expressed on immature B through early pre-B stage cells, thymocytes (except CD4/CD8 double positive thymocytes), peripheral T cells, and bone marrow stromal cells. It can be a useful marker for identifying memory and effector T cells and differentiation of Treg and conventional T cells.
Catalog No. NC0556866