Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Vibration Nylon Gloves with Grain Leather Tool Grip

Catalog No. 19400074 Shop All Impacto Protective Products Products
Click to view available options
Size:
This item is not returnable. View return policy
This item is not returnable. View return policy

VEP padding in the palm, thumb webbing, and index finger protects users from cumulative trauma such as bruising from constant tool gripping.

  • Fingerless leather glove with stretchy nylon back
  • Soft grain leather palm cover for longer wear
  • Impact and shock absorbing VEP padding in the palm, between the thumb, and on the index finger
  • Thumb web area is protected from cumulative trauma such as bruising from gripping tools
  • Provides protection from impact, vibration, and abrasion
  • Ideal for use with hand tools and power tools

Specifications

Color

Blue, Yellow

Disposable

Reusable

Hand

Specific

Material

Nylon Lycra

Product Type

Anti-impact Gloves

Size

X-Large

Sterility

Non-sterile

Finger Style

Fingerless

For Use With (Application)

Construction, DIY Home Improvement

Glove Cuff Style

Open

Palm Type

Padded

Series

502-20

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.