Learn More
STEMCELL Technologies Anti-Human CD56 Antibody, Clone HCD56, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60021
Encompass_Preferred
Clone HCD56 is a mouse monoclonal antibody that reacts with human CD56 (NCAM), a member of the immunoglobulin (Ig) super family containing 5 Ig-like domains and two fibronectin type-3 domains in its extracellular portion. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of CD56 (NCAM). The ~140 kDa isoform of CD56 is considered the prototypic cell surface marker on human NK cells. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ HLA Buffy Coat CD56 Positive Selection Kit (Catalog #18085HLA) and EasySep™ Human CD56 Positive Selection Kit (Catalog #18055); partial blocking may be observed, as well as EasySep™ HLA CD2 Positive Selection Kit (Catalog #18657HLA) and EasySep™ Human NK Cell Enrichment Kit (Catalog #19055). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5073
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.