Learn More
Fluidigm Corp Anti-Human CD192/CCR2 (K036C2), 100 tests
SDP
Supplier:  Fluidigm Corp 3153023B
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CD192, also known as chemokine receptor CCR2, is expressed mainly by inflammatory monocytes, and binds two ligands, CCL2 and CCL7. CCR2. Its ligands play a role in recruiting inflammatory monocytes into target tissues, where they differentiate into proinflammatory and anti-inflammatory macrophages and promote tissue injury and remodeling, respectively.
Catalog No. NC1776510