Learn More
Fluidigm Corp ANTI-HUMAN CD11C-147SM
SDP
Supplier:  Fluidigm Corp 3147008B
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CD11c is a transmembrane glycoprotein also known as integrin αX or CR4. CD11c is primarily expressed on dendritic cells, NK cells, a subset of intestinal intraepithelial lymphocytes (IEL), and some activated T cells. It has also been associated with hairy cell leukemias, acute nonlymphocytic leukemias, and some B cell chronic lymphocytic leukemias.
Catalog No. NC0790383