Learn More
Fluidigm Corp ANTI-HU CD127 -176YB
SDP
Supplier:  Fluidigm Corp 3176004B
Encompass_Preferred
CD127, also known as the IL-7 receptor α chain or IL-7α, is a 60–90 kDa type I transmembrane glycoprotein. It is a heterodimer consisting of IL-7Rα (CD127) and a common-γ chain receptor (CD132), which is shared with various cytokines including IL-2,-4,-9,-13,-15 and -21. CD127 is expressed on immature B through early pre-B stage cells, thymocytes (except CD4/CD8 double positive thymocytes), peripheral T cells, and bone marrow stromal cells. It can be a useful marker for identifying memory and effector T cells and differentiation of Treg and conventional T cells.
Catalog No. 50-467-017
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.