Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fas Mouse anti-Human, Clone: ZB4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05338
Description
Anti-Fas Antibody (human, neutralizing), clone ZB4 detects level of Fas and has been published and validated for use in western blot, flow cytometry, NEUT.Specifications
Fas | |
Monoclonal | |
Unconjugated | |
NM_152877 | |
Mouse | |
Protein A purfied | |
Primary | |
Stable for 1 year at from date of receipt. | |
IgG1 |
Flow Cytometry, Neutralization, Western Blot | |
ZB4 | |
Ammonium sulfate and purified mouse monoclonal IgG1 in PBS containing 50% glycerol. Store at −20°C. | |
FAS, OTTHUMP00000059646, LFG, FASTM, ALPS1A, TNFRSF6, CD95, FAS1, FAIM2, APT1, NMP35, APO-1 | |
Recombinant human Fas. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction