Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Dopamine D2S Receptor, Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ 324396100UL
Description
Specifically detects Dopamine D2S Receptor Clone: in Human, Rat samples, and it is validated for ELISA, Immunohistochemistry, Immuno Blotting
Specifications
Dopamine D2S Receptor | |
Polyclonal | |
Undiluted serum. | |
a synthetic (PLKEAARRC) peptide corresponding to amino acids 239-246 of human D2s dopamine receptor | |
RUO | |
Recognizes the ∽48kDa dopamine D25 receptor. Does not cross-react with other dopamine receptors. | |
2°C to 8°C | |
IgG |
ELISA, Immunoblot, Immunoblot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
100 μL | |
Primary | |
Human, Rat | |
Undiluted |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction