Learn More
Enzo Life Sciences Ansamitocin P-3 (1 mg). CAS: 66547-09-9
Supplier: Enzo Life Sciences BMLT1210001
Ansamitocin P-3 is a maytansine analog which displays potent cytotoxicity against the human solid tumor cell lines A-549 and HT-29. It shares the same binding site as rhizoxin and phomopsin A on porcine brain tubulin. Maytansine and its analogs cause extensive disassembly of the microtubule and totally prevent tubulin spiralization. Purity: ≥93% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.