Learn More
Enzo Life Sciences Angiotensin I ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900203
Encompass_Preferred
The Angiotensin I EIA kit is a colorimetric, competitive immunoassay kit with results in 3.5 hours. Sensitivity: 4.3 pg/ml (range 3.9-10,000 pg/ml). Use/Stability: All components of this kit, except the Standard (80‐2361) and Conjugate (80‐2363), should be stored at +4°C upon receipt. UniProt ID: P01019 (human). Assay Time: 3.5 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent . Content: Microtiter Plate, Conjugate, Antibody, Assay Buffer 16, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Streptavidin-HRP.
Catalog No. 50-200-6865
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.