Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amyris Oil W.I., Spectrum™ Chemical
Click to view available options
Quantity:
100 mL
500 g
500 mL
2.5 kg
50 lb.
Packaging:
Amber Glass Bottle
Drum
Specifications
Specifications
CAS Max % | 1 |
Appearance | Pale Yellow |
Packaging | Amber Glass Bottle |
Refractive Index | 1.495-1.525 |
Quantity | 500 mL |
Specific Gravity | 0.950 - 0.975 |
Grade | Reagent |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Share a Content Correction