Learn More
Enzo Life Sciences AMPIGENE dNTP Mix (500µl)
Supplier: Enzo Life Sciences ENZNUC1020500
AMPIGENE dNTP Mix is an ultrapure mix of deoxyribonucleotide triphosphates to use as building blocks for a wide variety of PCR applications. It can withstand multiple freeze-thaw cycles and shown to retain 95% dNTPs in triphosphate form after 5 weeks at room temperature. AMPIGENE dNTP Mix has been tested on many molecular biology applications including standard PCR, fast PCR, 1-step PCR, high-fidelity PCR, real-time PCR, lon-range PCR, MDA, RDA, LAMP, and DNA sequencing. Long Term Storage: -20°C. Applications: PCR.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.