Learn More
Enzo Life Sciences AMPIGENE cDNA Synthesis Kit (200 Reactions)
SDP
Supplier:  Enzo Life Sciences ENZKIT1060200
Encompass_Preferred
AMPIGENE cDNA Synthesis Kit uses the latest developments in reverse transcriptase (RTase) technology and buffer chemistry for efficient and accurate cDNA synthesis. The modified MMLV RTase is blended with RNase inhibitor to prevent degradation of RNA by contaminating RNase. The RTase is thermostable and extremely active. An optimized buffer system contains oligo(dT), random hexamers, enhancers, dNTPs and MgCl2, allowing efficient and sensitive cDNA synthesis.
Catalog No. 50-201-3813
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.