Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Amicon™ Pro Purification System with Ultra-0.5 Device, 10 Kda
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
$233.00
Specifications
Disposable | Single-use |
---|---|
MWCO | 10 kDa |
Sterility | Non-sterile |
Housing Material | Regenerated Cellulose (RC) |
Type | Centrifugal Filters |
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity & Availability | ||||
ACS501012
|
MilliporeSigma™
ACS501012 |
12 Amicon Ultra NMWCO 10 kDa filters, 12 Exchange devices, and 12 Collection tubes with cap | 12Pack |
Pack of 12 for $233.00
|
|
||||
ACS501024
|
MilliporeSigma™
ACS501024 |
24 Amicon Ultra NMWCO 10 KDa filters, 24 Exchange devices, and 24 Collection tubes with cap | 24Pack | N/A | N/A | N/A | |||
ACS501002
|
MilliporeSigma™
ACS501002 |
2 Amicon Ultra NMWCO 10 kDa filters, 2 Exchange devices, and 2 Collection tubes with cap | 2Pack | N/A | N/A | N/A | |||
Description
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.Specifications
Single-use | |
Non-sterile | |
Centrifugal Filters |
10 kDa | |
Regenerated Cellulose (RC) | |
12 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title