Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
American Educational Products Agar Powder
Supplier: American Educational Products 7CH001
This item is not returnable.
View return policy
Description
- Growth medium to culture microorganisms in the laboratory
- Convenient 3g size
- 3g units contain enough for approximately 1/4 in. of agar
- Growth will appear as dots of varying sizes on agar — incubate in warm location (around 98°F [36.6°C]) for faster growth
Specifications
Powder | |
Agar | |
Cultivation of microorganisms in the laboratory | |
3 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction