Learn More
Invitrogen™ Microcentrifuge Spin Columns
Sterile, RNase-free
Supplier: Invitrogen™ R50020
Description
The microcentrifuge spin columns included with the FastTrack™ 2.0 and Micro-FastTrack 2.0 Kits are available separately. The columns are made from polypropylene and are designed to enhance the speed and efficiency of polyA+ RNA isolation. The columns are sterilized and RNase-free.
DNA and RNA Purification and Analysis, RNA Extraction, mRNA Isolation
Specifications
0.45 micron cellulose acetate microcentrifuge spin columns and sterile microcentrifuge tubes. After opening, store microcentrifuge columns and tubes in a sterile container. | |
Spin Column | |
200 Columns |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.