Learn More
Accuflow Systems Sterile Empty Vial (SEV), 30 mL, 20 mm amber glass with silver cap - 25/pack
Supplier: Accuflow Systems SEV30AMB
ALK Sterile Empty Vials (SEV) feature Type 1 glass USP borosilicate, and a latex free stopper. ALK SEVs are produced at a cGMP FDA Manufacturer and USP tested for sterility and endotoxins. Certificate of Analysis available for all production lots and vials are aseptically assembled in an ISO 5 (class 100) cleanroom.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.