Learn More
Aluminum plate, alloy 6061, 9.53mm (0.375in) thick, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 042128RF
Description
Aluminum plate is a raw material for parts as diverse as automotive side panels, storage tank walls, architectural panels and signs. It is also used in manufacturing of fast ferries, yachts, navy or passenger vessels and small pleasure crafts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aluminum plate, Alloy 6061 | |
MFCD00214039 | |
Insoluble in water. | |
≈2315g/30x30cm |
9.53mm (0.375 in.) thick | |
30 x 30 cm | |
Odorless | |
Plate |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only