Learn More
Electron Microscopy Sciences Aluminum Oxide Powder, 5.0 µm 1 LB
Supplier:  Electron Microscopy Sciences 5036205
Encompass_Preferred
These are abrasive powders that are prepared from aluminum oxide, offering excellent coarse and intermediate polishing, with fast results.
Catalog No. 50-997-304
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.