Learn More
Thermo Scientific™ Alternate Cord and Plug Options for Ultra-Low Temperature Freezers
Meet your local requirements with Thermo Scientific™ Alternate Cord and Plug Options for Ultra-Low Temperature Freezers.
Supplier:  Thermo Scientific™ US230V15ATL
 View more versions of this product
Catalog No. 13-762-393
Explore all special offers
Add to Cart
Description
Description
Meet your local requirements with Thermo Scientific™ Alternate Cord and Plug Options for Ultra-Low Temperature Freezers. Options include an array of standard and special plugs for North America and international markets.
Specifications
Specifications
United States Twist Lock 208-230V/60Hz (Factory Installed)
Thermo Scientific™ TDE and TSX Series,Thermo Scientific™ TSU Series, HERAfreeze™ HFU-T, Revco™ UxF
NEMA 6-15 P
Twist Lock Plug
15 A
230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.