Learn More
JDM SCIENTIFIC LLC AlteCap™ Switch HT
Supplier:  JDM SCIENTIFIC LLC 404998
Encompass_Preferred
Semi-automated handheld decapper. With reduced torque for specific manufacturers tubes & caps. Electronic capping & decapping. Benchtop space saving design. Cap driver cassette for customer's tube type purchased separately. *For use with Switch cassette 404013 (fluidX) ONLY.
Catalog No. 50-271-9957
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.