Learn More
JDM SCIENTIFIC LLC AlteCap™ Switch Cassette 12/96 - Azenta Acoustic Sample Tube
Supplier:  JDM SCIENTIFIC LLC 404038
Encompass_Preferred
AlteCap™ Switch cap driver cassette - 12 channel. Compatible with 96 SBS rack format Azenta Acoustic Sample Tubes
Catalog No. 50-271-9981
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.