Learn More
AlphaProTech Critical Cover™ BarrierTech™ Gowns
Provides protection for the wearer from particles, fluids and light chemical splash
$512.00
Specifications
Cuff Style | Elastic |
---|---|
For Use With (Application) | Protection for the wearer from particles, fluids and light chemical splash |
Material | BarrierTech™ Fabric |
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
19-169-4038
|
AlphaProTech
GN225243 |
White | Universal |
Case of 50 for $512.00
|
|
||||
Description
- Offering a durable and fluid impervious solution for both your lab and/or production areas
- Low particle count for use in controlled environments
- Banded ties are attached to the neck and waist for easy and secure closure
- Serged seams and elastic wrists
- Available in two colors
Specifications
Elastic | |
Protection for the wearer from particles, fluids and light chemical splash | |
BarrierTech™ Fabric |
Spot an opportunity for improvement?
Share a Content Correction