Learn More
Enzo Life Sciences alpha-Latrotoxin (40µg). CAS: 65988-34-3
SDP
Supplier:  Enzo Life Sciences ALX630027C040
Encompass_Preferred
Causes massive neurotransmitter release from a wide variety of central and peripheral synaptic junctions of vertebrates using Ca2+-dependent and Ca2+-independent pathways. A useful pharmacological tool in the studies of synaptic vesicles exocytosis of different neutrotransmitters. Causes massive neurotransmitter release from a wide variety of central and peripheral synaptic junctions of vertebrates using Ca2+-dependent and Ca2+-independent pathways. A useful pharmacological tool in the studies of synaptic vesicles exocytosis of different neutrotransmitters. Purity: ≥97%. Formulation: Lyophilized. Source: Isolated from Latrodectus tredecimguttatus. Long Term Storage: -20°C.
Catalog No. 50-200-9609
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.