Learn More
eMolecules Medchem Express (US) | ALK-IN-27 | 100mg | HY-156467
Supplier: eMolecules 841055066
Medchem Express (US) | ALK-IN-27 | 100mg | HY-156467 | Purity: 97% | 2739866-40-9 | | MW: 452.92 NVL-655 is a selective inhibitor of anaplastic lymphoma kinase (ALK), with potential antineoplastic activity in the Ba/F3 xenograft model.