Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ IMPACT RPR™ Dispensing Bottle

For use with the IMPACT RPR™ product line.

Supplier:  Abbott™ 61C9

Catalog No. WA-61C9


Explore all special offers
Add to Cart

Description

Description

  • Classified as a standard test for syphilis by the Centers for Disease Control and Prevention (CDC)
  • Non-treponemal test for the serological detection of syphilis
  • Assay utilizes charcoal particle technology

Specifications

Specifications

Dispensing Bottle
Dynex DSX and DS2™ fully automated ELISA processing system
IMPACT RPR™
10 Pack
Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.