Learn More
Enzo Life Sciences Alectinib. hydrochloride (200mg). CAS: 1256589-74-8
SDP
Supplier:  Enzo Life Sciences ENZCHM2130200
Encompass_Preferred
Alectinib hydrochloride is a selective anaplastic lymphoma kinase (ALK) inhibitor leading to inhibition of tumor cell proliferation and induction of cell death.  Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3540
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.