Learn More
Andwin Scientific ALCOJET DETERGENT 4LBALCOJET
Supplier: Andwin Scientific 6051301
The Alcojet� Low Foaming Powdered Detergent is composed of complex phosphate, concentrated organic chlorine, sodium carbonate, and an alkylkoxylate. This product is ideal for use in the sanitation of health care instruments, laboratory ware, sampling apparatus, plate glass, pharmaceutical apparatus, cosmetics manufacturing equipment, restaurant equipment, wine glasses, counters, floors, sidewalks and driveways, metal castings, forgings and stampings, industrial parts, and tanks and reactors. Alcojet� can remove unwanted contaminants like soil, grit, grease, and many more particles. This product is sold in a quantity of 4 pounds.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.