Learn More
Eurofins Discoverx β-Alanine, 10 mg. (92-1227)
Supplier:  Eurofins Discoverx 921227
Encompass_Preferred
β-Alanine is a common non-essential beta amino acid formed by degradation of dihydrouracil and carnosine. Levels of β-alanine limit the amount of carnosine, and it has been used frequently as a workout supplement to increase carnosine concentrations in muscle. This assists in building lean muscle mass, and can also improve physical functioning and performance. Acts as an agonist for MRGPRD. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9750
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.