Learn More
Eurofins Discoverx Akt1/PKBα, active, 250 µg. (14-276M)
Supplier:  Eurofins Discoverx 14276M
Encompass_Preferred
N-terminal 6xHis-tagged recombinant, human full length Akt1. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8740
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.