Learn More
MP Biomedicals™ AIN-76A Semipurified Diet
AIN-76A Semipurified Diet is intended for growth and maintenance of animals during the first year of life.
$983.00 - $3714.00
Products 3
Catalog Number Mfr. No. dimQuantity Price Quantity & Availability  
Catalog Number Mfr. No. dimQuantity Price Quantity & Availability  
02-960-09710
View Documents
MP Biomedicals™
0296009710
Each for $983.00
Add to Cart
02-960-09720
View Documents
MP Biomedicals™
0296009720
Each for $1,711.00
Add to Cart
02-960-09750
View Documents
MP Biomedicals™
0296009750
Each for $3,714.00
Add to Cart
Description
Description
  • Contains casein purified high nitrogen, DL-methionine, sucrose, corn starch, Alphacel (non-nutritive bulk), corn oil, choline bitartrate, AIN mineral mix, and a 10-fold increase in the amount of vitamin K source to 500 μg/kg.
Videos
SDS
Documents
Documents
Product Certifications
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.