Learn More
Enzo Life Sciences AG-879 (5mg). CAS: 148741-30-4
SDP
Supplier:  Enzo Life Sciences BMLEI2580005
Encompass_Preferred
AG-879, a member of the tyrphostin family of tyrosine kinase inhibitors, inhibits NGF receptor (pp140c-trk) autophosphorylation selectively with no inhibition of EGF or PDGF receptor phosphorylation (IC50=10 µM). AG-879 inhibits NGF-induced neurite outgrowth in PC12 cells. Purity: ≥99% (HPLC). Solubility: Soluble in 100% ethanol (25mg/ml) or DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0638
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.