Learn More
Electron Microscopy Sciences AFM/STM 90° Mount w/Spring Clip
Supplier:  Electron Microscopy Sciences 7400002
Encompass_Preferred
These mounts and sample holders make it easy to clamp and position cross sectional samples for AFM imaging. They're easy to attach to the magnetic sample holder of the AFM system. Compatible with most AFM systems on the market. Made from high quality magnetic stainless steel.
Catalog No. 50-193-0070
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.