Learn More
Agilent Technologies AFFINITYSCRIPT MULT TEMP TRANS
Supplier:  Agilent Technologies 600107
Encompass
Our AffinityScript Multiple Temperature Reverse Transcriptase is engineered to be highly thermostable, allowing you to reverse transcribe at your preferred reaction temperature.
Catalog No. NC9519419
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.