Learn More
Cosmo Bio Usa Inc Advanced A-PAP Pen Liquid-Blocker Mini: PAP Pens have been improved! The new formulation of Advanced A-PAP Pens is more easily removed with xylene.
SDP
Supplier:  Cosmo Bio Usa Inc DAIAPAPM
Encompass
PAP Pens have been improved! The new formulation of Advanced A-PAP Pens is more easily removed with xylene. Additionally, water repellency and quick-drying are slightly faster than previous liquid blockers. The protocol, volume of the tube, and the diameter of the tip remain the same. Ready-to-use hydrophobic barrier pen designed for immunohistochemistry applications. This ready-to-use hydrophobic barrier pen is designed for immunohistochemistry applications. The surface tension provided by the circle, square, or any other shapes drawn to t tissue sections with the pen, ensures that only the amount of antibody solution needed for the su cient reaction will be applied. The pen is stable up to 129 °C in microwave heat. The PAP Pen is an effective tool for immunostaining procedures by the Peroxidase-Antiperoxidase (PAP) method, ABC method, LSAB method, polymeric method, immunofluorescence method, ASD method, Enzyme method and Frozen Section method. This replaces DAI-PAP-S-M.
Catalog No. NC2063118
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.