Learn More
MilliporeSigma™ Supelco™ Adsorbent Scrapers
For Rapid Removal of Adsorbent from TLC Plates
Supplier:  MilliporeSigma™ Supelco™ Z2652681EA
 View more versions of this product
Catalog No. 11-101-1797
Description
Description
  • Flat 13 mm steel blade tapered to a sharp edge
  • Ideal for rapid removal of adsorbent from TLC plates
  • Aluminum handle
  • Replaceable blade
Specifications
Specifications
Aluminum
1 Ea.
Adsorbent Scrapers
Rapid Removal of Adsorbent From TLC Plates
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.