Learn More
Enzo Life Sciences Adipogenesis assay kit (5x24wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1030005
Encompass_Preferred
The Adipogenesis Assay Kit provides all the reagents needed to induce and detect adipogenesis. Optimized Differentiation, Insulin, and Enhancer solutions ensure maximal adipocyte induction. The Adipogenesis Dye allows for visual confirmation of differentiation and can also be easily extracted allowing measurement using a spectrophotometer or microplate reader.  Applications: Colorimetric detection .
Catalog No. 50-201-3807
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.