Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenovirus Hexon Antibody (3G0), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065051F
Description
Adenovirus Hexon Monoclonal antibody specifically detects Adenovirus Hexon in Adenovirus, Virus samples. It is validated for ELISA, ImmunofluorescenceSpecifications
Adenovirus Hexon | |
Monoclonal | |
FITC | |
Hexon protein; Late protein 2; PII | |
ATCC VR5 virus | |
0.1 mL | |
Virology Bacteria and Parasites | |
Adenovirus, Virus | |
Purified |
ELISA, Immunofluorescence | |
3G0 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction