Learn More
Chemglass Life Sciences Adapter, Distilling/Splitting, 29/42, Angled, 0-4mm Valve, 1/2" Compression Fitting, Process Reactor
SDP
Supplier:  Chemglass Life Sciences CG1235B29
Encompass_Preferred
Adapter, Distilling/Splitting, 29/42, Angled, 0-4mm Valve, 1/2" Compression Fitting, Process Reactor. Compact and simple distilling/splitter adapter for use with Chemglass process reactors lids that have a 10° angled side neck (CG-1946-B). Vapor flows upward freely through the large vapors holes to the condenser. Distillate flow is directed using the valve to either the receiving flask or back to the reactor. Control valve is either a 0-4mm or a 0-12mm PTFE with a 1/2” OD distillate takeoff for use with CG-1975 compression fittings and TFE tubing. Plugs are supplied complete with CG-309 Perfluoro o-rings.
Catalog No. 50-143-594
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.